Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxybenzimidazole 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
615-16-7
C7H6N2O
134.138
MFCD00127894
SILNNFMWIMZVEQ-UHFFFAOYSA-N
2-hydroxybenzimidazole,2-benzimidazolol,2-benzimidazolinone,o-phenyleneurea,2-benzimidazolone,2-oxobenzimidazole,2 3h-benzimidazolone,1,3-dihydro-2h-benzimidazol-2-one,2h-benzimidazol-2-one, 1,3-dihydro,benzamidazole-2 3h-one
11985
1,3-dihydrobenzimidazol-2-one
C1=CC=C2C(=C1)NC(=O)N2
615-16-7
C7H6N2O
134.138
MFCD00127894
SILNNFMWIMZVEQ-UHFFFAOYSA-N
2-hydroxybenzimidazole,2-benzimidazolol,2-benzimidazolinone,o-phenyleneurea,2-benzimidazolone,2-oxobenzimidazole,2 3h-benzimidazolone,1,3-dihydro-2h-benzimidazol-2-one,2h-benzimidazol-2-one, 1,3-dihydro,benzamidazole-2 3h-one
11985
1,3-dihydrobenzimidazol-2-one
C1=CC=C2C(=C1)NC(=O)N2
Quantity 25g
Melting Point 314.0°C to 316.0°C
Infrared Spectrum Authentic
Color Brown to White
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 134.14
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Hydroxybenzimidazole, 97%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-412-4

RTECSNumber : DE1625000

TSCA : TSCA