Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxybenzyl)triphenylphosphonium bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
70340-04-4
C25H22BrOP
449.328
MFCD00011902
FKKZGQXMWVGPMH-UHFFFAOYSA-N
2-hydroxybenzyl triphenylphosphonium bromide,2-hydroxyphenyl methyl-triphenylphosphanium bromide,2-hydroxyphenyl methyl triphenylphosphanium bromide,phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide,acmc-209oe4,2-hydroxybenzyltriphenylphosphonium bromide,2-hydroxybenzyl triphenylphosphoniumbromide,2-hydroxybenzyl triphenyl phosphonium bromide,ortho hydroxybenzyltriphenyl-phosphonium bromide,2-hydroxyphenyl methyl-triphenylphosphonium bromide
2724142
(2-hydroxyphenyl)methyl-triphenylphosphanium;bromide
C1=CC=C(C=C1)[P+](CC2=CC=CC=C2O)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
70340-04-4
C25H22BrOP
449.328
MFCD00011902
FKKZGQXMWVGPMH-UHFFFAOYSA-N
2-hydroxybenzyl triphenylphosphonium bromide,2-hydroxyphenyl methyl-triphenylphosphanium bromide,2-hydroxyphenyl methyl triphenylphosphanium bromide,phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide,acmc-209oe4,2-hydroxybenzyltriphenylphosphonium bromide,2-hydroxybenzyl triphenylphosphoniumbromide,2-hydroxybenzyl triphenyl phosphonium bromide,ortho hydroxybenzyltriphenyl-phosphonium bromide,2-hydroxyphenyl methyl-triphenylphosphonium bromide
2724142
(2-hydroxyphenyl)methyl-triphenylphosphanium;bromide
C1=CC=C(C=C1)[P+](CC2=CC=CC=C2O)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
For C-C Bond Formation (Olefination)
Packaging Glass bottle
Quantity 10g
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 240°C to 244°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H4CH2P(C6H5)3Br
Formula Weight 449.33
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material (2-Hydroxybenzyl)triphenylphosphonium bromide, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 274-574-8

TSCA : TSCA