Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl acrylate, 97%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5mL
250mL
1L
2.5L
Packaging:
Glass bottle
818-61-1,150-76-5
C5H8O3
116.12
MFCD00002865
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl acrylate,hydroxyethyl acrylate,2-propenoic acid, 2-hydroxyethyl ester,ethylene glycol monoacrylate,bisomer 2hea,acrylic acid 2-hydroxyethyl ester,2-acryloyloxy ethanol,ethylene glycol, acrylate,acrylic acid, 2-hydroxyethyl ester,2-hydroxyethylacrylate
13165
2-hydroxyethyl prop-2-enoate
C=CC(=O)OCCO
818-61-1,150-76-5
C5H8O3
116.12
MFCD00002865
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl acrylate,hydroxyethyl acrylate,2-propenoic acid, 2-hydroxyethyl ester,ethylene glycol monoacrylate,bisomer 2hea,acrylic acid 2-hydroxyethyl ester,2-acryloyloxy ethanol,ethylene glycol, acrylate,acrylic acid, 2-hydroxyethyl ester,2-hydroxyethylacrylate
13165
2-hydroxyethyl prop-2-enoate
C=CC(=O)OCCO
Melting Point -60°C
Boiling Point 210°C to 215°C
Flash Point 99°C
Viscosity 9 mPa.s (20°C)
Specific Gravity 1.106
Refractive Index 1.4490 to 1.451
Infrared Spectrum Authentic
Quantity 2.5L
Packaging Glass bottle
Color Yellow to Brown
Stabilizer 200 to 450ppm MEHQ
Density 1.1060g/mL
Acidity 4mg KOH/g max.
Water 0.15% max.
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: soluble in water
Formula Weight 116.12
Physical Form Oily Liquid
Percent Purity 97%
Chemical Name or Material 2-Hydroxyethyl acrylate, Stabilized
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-454-9