Learn More
(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals
Description
(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -70°C |
Density | 1.121 g/mL |
Boiling Point | 218°C to 220°C |
Flash Point | 73°C (163°F) |
Refractive Index | 1.493 |
Quantity | 50 g |
UN Number | UN2810 |
Solubility Information | Soluble in water,alcohols and glycols. |
Formula Weight | 76.1 |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H227-H301-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H227-H315-H319-H335 Toxic if swallowed. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 203-711-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.