Alfa Aesar™ 2-Hydroxyethyl methacrylate, 97%, stab. with ca 500ppm 4-methoxyphenol

Manufacturer: Alfa Aesar™ B2426036

Quantity 500g

 View more versions of this product

Catalog No. AAB2426036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 868-77-9
    Molecular Formula CH2=CH(CH3)CO2CH2CH2OH
    Formula Weight 130.14
    MDL Number MFCD00002863
    Beilstein 1071583
    Synonym Ethylene glycol monomethacrylate; Methacrylic acid 2-hydroxyethyl ester
    Chemical Name or Material 2-Hydroxyethyl methacrylate, stabilized with ≈500ppm 4-methoxyphenol

    Specifications

    Formula Weight 130.14
    TSCA TSCA
    CAS 868-77-9
    Beilstein 1071583
    Synonym Ethylene glycol monomethacrylate; Methacrylic acid 2-hydroxyethyl ester
    Health Hazard 2 GHS H Statement
    H315-H319-H317
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P362-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    RTECS Number OZ4725000
    Flash Point 101°C (213°F)
    Odor Ester-like
    Melting Point −12°C
    Quantity 500g
    Molecular Formula CH2=CH(CH3)CO2CH2CH2OH
    MDL Number MFCD00002863
    Health Hazard 1 Warning
    Boiling Point 250°C
    Chemical Name or Material 2-Hydroxyethyl methacrylate, stabilized with ≈500ppm 4-methoxyphenol
    Assay 97%

    Description

    Applications
    2-Hydroxyethyl methacrylate is used in UV-curable inks and coatings. It is also used in adhesives, artificial nails, dental materials and lacquers. In dentistry, it is one of the main volatile acrylates along with methyl methacrylate. Further, it is used as a monomer in the synthesis of polymers for dental prosthetics and for geotechnical grouting in construction work.

    Solubility
    Slightly miscible with hot water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H317
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P362-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit