Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl stearate, ACROS Organics™

Click to view available options
Quantity:
100g
250g
Packaging:
Glass bottle
111-60-4
C20H40O3
328.537
MFCD00051465
RFVNOJDQRGSOEL-UHFFFAOYSA-N
glycol stearate,cremophor a,2-hydroxyethyl stearate,polyethylene glycol monostearate,ethylene glycol monostearate,cerasynt m,clearate g,cerasynt mn,cithrol ps,polyoxyl 40 stearate
24762
2-hydroxyethyl octadecanoate
CCCCCCCCCCCCCCCCCC(=O)OCCO
111-60-4
C20H40O3
328.537
MFCD00051465
RFVNOJDQRGSOEL-UHFFFAOYSA-N
glycol stearate,cremophor a,2-hydroxyethyl stearate,polyethylene glycol monostearate,ethylene glycol monostearate,cerasynt m,clearate g,cerasynt mn,cithrol ps,polyoxyl 40 stearate
24762
2-hydroxyethyl octadecanoate
CCCCCCCCCCCCCCCCCC(=O)OCCO
Packaging Glass bottle
Melting Point 55°C to 60°C
Quantity 250g
Infrared Spectrum Authentic
Color White to Yellow
Concentration or Composition (by Analyte or Components) compound is a mixture of various chain-length esters and di(ethylene glycol) ester
Formula Weight 328.53
Physical Form Flakes
Grade Technical
Chemical Name or Material 2-Hydroxyethyl stearate

EINECSNumber : 203-886-9

RTECSNumber : WI4100000

TSCA : TSCA