Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (2-Hydroxyethyl)trimethylammonium dimethyl phosphate, 95%

Manufacturer:  Alfa Aesar™ H2760918

Quantity 50g

 View more versions of this product

Catalog No. AAH2760918


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

118978-98-6
(HOCH2CH2)N(CH3)3·(CH3O)2PO2
MFCD09953490
C7H20NO5P
229.21
(2-Hydroxyethyl)trimethylammonium dimethyl phosphate

Specifications

40° to 41°C
118978-98-6
(HOCH2CH2)N(CH3)3·(CH3O)2PO2
MFCD09953490
95%
50g
C7H20NO5P
229.21
(2-Hydroxyethyl)trimethylammonium dimethyl phosphate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit