Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyledisulfide, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1892-29-1
C4H10O2S2
154.242
MFCD00002906
KYNFOMQIXZUKRK-UHFFFAOYSA-N
2-hydroxyethyl disulfide,2,2'-dithiodiethanol,bis 2-hydroxyethyl disulfide,ethanol, 2,2'-dithiobis,hydroxyethyl disulfide,diethanol disulfide,2,2'-dithiobisethanol,2-hydroxyethanedisulfide,usaf th-9,2,2'-disulfanediyldiethanol
15906
CHEBI:43136
2-(2-hydroxyethyldisulfanyl)ethanol
C(CSSCCO)O
1892-29-1
C4H10O2S2
154.242
MFCD00002906
KYNFOMQIXZUKRK-UHFFFAOYSA-N
2-hydroxyethyl disulfide,2,2'-dithiodiethanol,bis 2-hydroxyethyl disulfide,ethanol, 2,2'-dithiobis,hydroxyethyl disulfide,diethanol disulfide,2,2'-dithiobisethanol,2-hydroxyethanedisulfide,usaf th-9,2,2'-disulfanediyldiethanol
15906
CHEBI:43136
2-(2-hydroxyethyldisulfanyl)ethanol
C(CSSCCO)O
Color Colorless to Yellow
Water 10% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Density 1.2877g/mL
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula (HOCH2CH2)2S2
Beilstein 01, 471
Formula Weight 154.25
Physical Form Viscous Liquid
Percent Purity ≥88%
Chemical Name or Material 2-Hydroxyethyledisulfide
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger