Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl salicylate, 98+%, Acros Organics

Click to view available options
Quantity:
50mL
250mL
1L
Packaging:
Glass bottle
87-28-5
C9H10O4
182.18
MFCD00002862
LVYLCBNXHHHPSB-UHFFFAOYSA-N
2-hydroxyethyl salicylate,glycol salicylate,ethylene glycol monosalicylate,espirosal,glycol monosalicylate,rheumacyl,sarocol,spirosal,glysal,phlogont
6880
CHEBI:86541
2-hydroxyethyl 2-hydroxybenzoate
C1=CC=C(C(=C1)C(=O)OCCO)O
87-28-5
C9H10O4
182.18
MFCD00002862
LVYLCBNXHHHPSB-UHFFFAOYSA-N
2-hydroxyethyl salicylate,glycol salicylate,ethylene glycol monosalicylate,espirosal,glycol monosalicylate,rheumacyl,sarocol,spirosal,glysal,phlogont
6880
CHEBI:86541
2-hydroxyethyl 2-hydroxybenzoate
C1=CC=C(C(=C1)C(=O)OCCO)O
Infrared Spectrum Authentic
Boiling Point 172°C (12.0mmHg)
Flash Point 174°C
Melting Point 25°C
Specific Gravity 1.25
Density 1.2500g/mL
Color Colorless
Quantity 50mL
Packaging Glass bottle
Assay Percent Range 98% min. (GC)
Linear Formula 2-(HO)C6H4CO2CH2CH2OH
Beilstein 10, 81
Merck Index 15, 4535
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: very soluble in alcohol,benzene,chloroform,,ether
Formula Weight 182.18
Physical Form Oily Liquid or Low Melting Solid
Percent Purity 98+%
Chemical Name or Material 2-Hydroxyethyl salicylate
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-737-2