Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Hydroxyflavanone, 98%

Click to view available options
Quantity:
1g
5g
25g
35244-11-2
C15H12O3
MFCD00017702
2r-2-2-hydroxyphenyl-2,3-dihydrochromen-4-one,2r-2-2-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one,2r-2-2-hydroxyphenyl-3,4-dihydro-2h-1-benzopyran-4-one,2r-2-2-hydroxyphenyl-2,3-dihydro-1-benzopyran-4-one
35244-11-2
C15H12O3
MFCD00017702
2r-2-2-hydroxyphenyl-2,3-dihydrochromen-4-one,2r-2-2-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one,2r-2-2-hydroxyphenyl-3,4-dihydro-2h-1-benzopyran-4-one,2r-2-2-hydroxyphenyl-2,3-dihydro-1-benzopyran-4-one
Quantity 1g
Formula Weight 240.25
Percent Purity 98%
Chemical Name or Material 2'-Hydroxyflavanone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning