Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyisobutyric acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Plastic bottle
594-61-6
C4H8O3
104.11
MFCD00004459
BWLBGMIXKSTLSX-UHFFFAOYSA-N
2-hydroxyisobutyric acid,2-methyllactic acid,acetonic acid,2-hydroxy-2-methylpropionic acid,alpha-hydroxyisobutyric acid,hydroxydimethylacetic acid,acetonate,propanoic acid, 2-hydroxy-2-methyl,alpha-hydroxyisobutanoic acid,2-hydroxyisobutyricacid
11671
CHEBI:50129
2-hydroxy-2-methylpropanoic acid
CC(C)(C(=O)O)O
594-61-6
C4H8O3
104.11
MFCD00004459
BWLBGMIXKSTLSX-UHFFFAOYSA-N
2-hydroxyisobutyric acid,2-methyllactic acid,acetonic acid,2-hydroxy-2-methylpropionic acid,alpha-hydroxyisobutyric acid,hydroxydimethylacetic acid,acetonate,propanoic acid, 2-hydroxy-2-methyl,alpha-hydroxyisobutanoic acid,2-hydroxyisobutyricacid
11671
CHEBI:50129
2-hydroxy-2-methylpropanoic acid
CC(C)(C(=O)O)O
Melting Point 77°C to 81°C
Boiling Point 84°C (1.5mmHg)
Quantity 1kg
Packaging Plastic bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2C(OH)CO2H
Beilstein 03, 313
Solubility Information Solubility in water: 1900g/L (22°C). Other solubilities: soluble in methanol,diethyl ether and hot benzene,sparingly soluble in cold benzene
Formula Weight 104.11
Percent Purity 98%
Chemical Name or Material 2-Hydroxyisobutyric acid, 98%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 209-848-8

TSCA : TSCA