Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxymethyl-15-crown-5, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
75507-25-4
C11H22O6
250.291
MFCD00192215
YHIQMMGCRYKJLB-UHFFFAOYSA-N
1,4,7,10,13-pentaoxacyclopentadecane-2-methanol,2-hydroxymethyl-15-crown-5,2-hydroxymethyl-15-crown 5-ether,1,4,7,10,13-pentaoxacyclopentadecan-2-ylmethanol,hydroxy-15-crown-5,2-hydroxymethyl-15-crown5-ether,2-hydroxymethyl-15-crown,2,5,8,11,14-pentaoxacyclopentadecyl methan-1-ol,1,4,7,10,13-pentaoxacyclopentadec-2-yl-methanol,1,4,7,10,13-pentaoxacyclopentadecan-2-yl methanol
2733744
1,4,7,10,13-pentaoxacyclopentadec-2-ylmethanol
C1COCCOCC(OCCOCCO1)CO
75507-25-4
C11H22O6
250.291
MFCD00192215
YHIQMMGCRYKJLB-UHFFFAOYSA-N
1,4,7,10,13-pentaoxacyclopentadecane-2-methanol,2-hydroxymethyl-15-crown-5,2-hydroxymethyl-15-crown 5-ether,1,4,7,10,13-pentaoxacyclopentadecan-2-ylmethanol,hydroxy-15-crown-5,2-hydroxymethyl-15-crown5-ether,2-hydroxymethyl-15-crown,2,5,8,11,14-pentaoxacyclopentadecyl methan-1-ol,1,4,7,10,13-pentaoxacyclopentadec-2-yl-methanol,1,4,7,10,13-pentaoxacyclopentadecan-2-yl methanol
2733744
1,4,7,10,13-pentaoxacyclopentadec-2-ylmethanol
C1COCCOCC(OCCOCCO1)CO
Chelation/Complexation reagent
Density 1.1g/mL
Packaging Glass bottle
Refractive Index 1.4780 to 1.4800 (20°C, 589nm)
Quantity 1g
Infrared Spectrum Authentic
Color Colorless
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 250.29
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 2-Hydroxymethyl-15-crown-5
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.