Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Hydroxymethyl)anthraquinone, 97%, Acros Organics

Click to view available options
Quantity:
1g
17241-59-7
C15H10O3
238.242
MFCD00001236
JYKHAJGLEVKEAA-UHFFFAOYSA-N
2-hydroxymethyl anthraquinone,2-hydroxymethylanthraquinone,2-hydroxymethyl anthracene-9,10-dione,anthraquinone-2-methanol,ccris 3522,9,10-anthracenedione, 2-hydroxymethyl,anthraquinone, 2-hydroxymethyl,2-hydroxymethyl-9,10-anthracenedione,2-hydroxymethyl-9,10-anthraquinone,2-hydroxymethyl anthraouinone
87014
CHEBI:28649
2-(hydroxymethyl)anthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)CO
17241-59-7
C15H10O3
238.242
MFCD00001236
JYKHAJGLEVKEAA-UHFFFAOYSA-N
2-hydroxymethyl anthraquinone,2-hydroxymethylanthraquinone,2-hydroxymethyl anthracene-9,10-dione,anthraquinone-2-methanol,ccris 3522,9,10-anthracenedione, 2-hydroxymethyl,anthraquinone, 2-hydroxymethyl,2-hydroxymethyl-9,10-anthracenedione,2-hydroxymethyl-9,10-anthraquinone,2-hydroxymethyl anthraouinone
87014
CHEBI:28649
2-(hydroxymethyl)anthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)CO
Quantity 1g
Color Yellow to Brown
Melting Point 192°C to 197°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 08, III, 2940
Fieser 08,252; 02,369; 04,432; 05,585; 06,518; 15,119; 16,120; 17,105
Formula Weight 238.24
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-(Hydroxymethyl)anthraquinone, 97%
Show More Show Less

LIGHT SENSITIVE

EINECSNumber : 241-274-3

RTECSNumber : CB7600000

TSCA : TSCA