Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Hydroxymethyl)pyridine 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
586-98-1
C6H7NO
109.13
MFCD00006348
SHNUBALDGXWUJI-UHFFFAOYSA-N
2-pyridinemethanol,2-hydroxymethyl pyridine,piconol,2-pyridinylmethanol,2-pyridylmethanol,2-pyridylcarbinol,pyridine-2-methanol,pyridine-2-carbinol,alpha-picolyl alcohol,pyridin-2-yl methanol
11474
pyridin-2-ylmethanol
C1=CC=NC(=C1)CO
586-98-1
C6H7NO
109.13
MFCD00006348
SHNUBALDGXWUJI-UHFFFAOYSA-N
2-pyridinemethanol,2-hydroxymethyl pyridine,piconol,2-pyridinylmethanol,2-pyridylmethanol,2-pyridylcarbinol,pyridine-2-methanol,pyridine-2-carbinol,alpha-picolyl alcohol,pyridin-2-yl methanol
11474
pyridin-2-ylmethanol
C1=CC=NC(=C1)CO
Packaging Glass bottle
Water 0.4% max. (K.F.)
Flash Point 117°C
Infrared Spectrum Authentic
Specific Gravity 1.13
Refractive Index 1.5410 to 1.5440
Quantity 100g
Color Brown or Colorless to Yellow
Density 1.1300g/mL
Melting Point 5.0°C
Boiling Point 220.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H4N)CH2OH
Beilstein 21, I, 203
Solubility Information Solubility in water: soluble
Formula Weight 109.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-(Hydroxymethyl)pyridine, 98%
Show More Show Less

GHS H StatementCauses serious eye irritation.Causes skin irritation.

GHS P StatementIF ON SKIN: Wash with plenty of soap and water.Call a POISON CENTER or doctor/physician if you feel unwell.If eye irritation persists: Get medical advice/attention.Wear protective gloves/protective clothing/eye protection/face protection.

;GHS P StatementIF ON SKIN: Wash with plenty of soap and water.Call a POISON CENTER or doctor/physician if you feel unwell.If eye irritation persists: Get medical advice/attention.Wear protective gloves/protective clothing/eye protection/fa

EINECSNumber : 209-592-7

RTECSNumber : UT4689000

TSCA : TSCA