Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
89466-08-0
C6H7BO3
137.93
YDMRDHQUQIVWBE-UHFFFAOYSA-N
2-hydroxyphenyl boronic acid,2-hydroxybenzeneboronic acid,o-hydroxyphenylboronic acid,2-boronophenol,2-hydroxyphenyl boranediol,2-hydroxyphenylboronicacid,boronic acid, 2-hydroxyphenyl,pubchem9522,2-hydroxyphenylboronic,acmc-209r1d
2773454
(2-hydroxyphenyl)boronic acid
B(C1=CC=CC=C1O)(O)O
89466-08-0
C6H7BO3
137.93
YDMRDHQUQIVWBE-UHFFFAOYSA-N
2-hydroxyphenyl boronic acid,2-hydroxybenzeneboronic acid,o-hydroxyphenylboronic acid,2-boronophenol,2-hydroxyphenyl boranediol,2-hydroxyphenylboronicacid,boronic acid, 2-hydroxyphenyl,pubchem9522,2-hydroxyphenylboronic,acmc-209r1d
2773454
(2-hydroxyphenyl)boronic acid
B(C1=CC=CC=C1O)(O)O
Melting Point 173.0°C to 174.0°C
Infrared Spectrum Authentic
Color White
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Linear Formula HOC6H4B(OH)2
Solubility Information Solubility in water: insoluble
Formula Weight 137.93
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Hydroxyphenylboronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning