Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480

Manufacturer:  Alfa Aesar™ H3113322

Quantity 100g

 View more versions of this product

Catalog No. AAH3113322


  Check Availability
Add to cart

Specifications

Description

Applications
2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
Description & Specifications

Chemical Identifiers

128446-35-5
≈1250 to 1480 d
MFCD00069372
(2-Hydroxypropyl)-beta-cyclodextrin

Specifications

305°C
128446-35-5
MFCD00069372
100g
≈1250 to 1480 d
(2-Hydroxypropyl)-beta-cyclodextrin

Safety and Handling

SDS & Safety Documents

RTECSNumber : GU2293332

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit