Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxypropyl)-beta-cyclodextrin, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Fiber Drum
128446-35-5
128446-35-5
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 1
Grade Reagent

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.