Alfa Aesar™ 2-Hydroxypropyl methacrylate

Suitable for embedding and microscopy

Manufacturer: Alfa Aesar™ 42116AO

Quantity 450mL

 View more versions of this product

Catalog No. AA42116AO

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 27813-02-1
    Formula Weight 144.17
    MDL Number MFCD00004536
    Chemical Name or Material 2-Hydroxypropyl methacrylate

    Specifications

    Formula Weight 144.17
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00004536
    Health Hazard 2 GHS H Statement
    H319-H317
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Density 1.078g/mL
    Boiling Point 205° to 209°C
    TSCA TSCA
    Physical Form Liquid
    CAS 27813-02-1
    Health Hazard 1 Warning
    Quantity 450mL
    Melting Point 70°C (1mmHg)
    Chemical Name or Material 2-Hydroxypropyl methacrylate

    GHS H Statement
    H319-H317
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit