Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxypyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
Plastic bottle
142-08-5
C5H5NO
95.1
MFCD00006268
UBQKCCHYAOITMY-UHFFFAOYSA-N
2-hydroxypyridine,pyridin-2-ol,2-pyridinol,2-pyridone,pyridin-2 1h-one,2 1h-pyridinone,2-pyridinone,2-oxopyridine,2 1h-pyridone,alpha-pyridone
8871
CHEBI:16540
1H-pyridin-2-one
C1=CC(=O)NC=C1
142-08-5
C5H5NO
95.1
MFCD00006268
UBQKCCHYAOITMY-UHFFFAOYSA-N
2-hydroxypyridine,pyridin-2-ol,2-pyridinol,2-pyridone,pyridin-2 1h-one,2 1h-pyridinone,2-pyridinone,2-oxopyridine,2 1h-pyridone,alpha-pyridone
8871
CHEBI:16540
1H-pyridin-2-one
C1=CC(=O)NC=C1
Color Undesignated
Melting Point 102.0°C to 109.0°C
Packaging Plastic bottle
Flash Point 210°C
Boiling Point 280.0°C to 281.0°C
Quantity 250g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 21, 43
Fieser 03,157
Solubility Information Solubility in water: soluble in water. Other solubilities: slightly soluble in chloroform and alcohol, practically insoluble in ether and benzene
Formula Weight 95.1
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Hydroxypyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger