Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxypyrimidine Hydrochloride 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
38353-09-2
C4H4N2O·ClH
132.55
MFCD00012781
IAJINJSFYTZPEJ-UHFFFAOYSA-N
2-hydroxypyrimidine hydrochloride,pyrimidin-2-ol hydrochloride,2 1h-pyrimidinone, monohydrochloride,2-hydroxypyrimidine hcl,2-pyrimidinone hydrochloride,pyrimidin-2 1h-one hydrochloride,2-hydroxypyrimidinehydrochloride,pyrimidin-2-one monohydrochloride,1,2-dihydropyrimidin-2-one hydrochloride,pyrimidin-2-one hydrochloride
122774
1H-pyrimidin-2-one;hydrochloride
C1=CNC(=O)N=C1.Cl
38353-09-2
C4H4N2O·ClH
132.55
MFCD00012781
IAJINJSFYTZPEJ-UHFFFAOYSA-N
2-hydroxypyrimidine hydrochloride,pyrimidin-2-ol hydrochloride,2 1h-pyrimidinone, monohydrochloride,2-hydroxypyrimidine hcl,2-pyrimidinone hydrochloride,pyrimidin-2 1h-one hydrochloride,2-hydroxypyrimidinehydrochloride,pyrimidin-2-one monohydrochloride,1,2-dihydropyrimidin-2-one hydrochloride,pyrimidin-2-one hydrochloride
122774
1H-pyrimidin-2-one;hydrochloride
C1=CNC(=O)N=C1.Cl
Melting Point 203°C to 205°C
Color Beige to Yellow
Packaging Glass bottle
Quantity 100g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 24, I, 231
Formula Weight 132.55
Physical Form Crystalline Solid
Percent Purity ≥97.5%
Chemical Name or Material 2-Hydroxypyrimidine hydrochloride, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 253-897-8

TSCA : TSCA