Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyquinoline 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
25g
Packaging:
Glass bottle
59-31-4
C9H7NO
145.161
MFCD00006743
LISFMEBWQUVKPJ-UHFFFAOYSA-N
2-hydroxyquinoline,2-quinolinol,quinolin-2-ol,carbostyril,2 1h-quinolinone,2-quinolone,quinolin-2 1h-one,quinolinol,quinolinone,alpha-quinolone
6038
CHEBI:18289
1H-quinolin-2-one
C1=CC=C2C(=C1)C=CC(=O)N2
59-31-4
C9H7NO
145.161
MFCD00006743
LISFMEBWQUVKPJ-UHFFFAOYSA-N
2-hydroxyquinoline,2-quinolinol,quinolin-2-ol,carbostyril,2 1h-quinolinone,2-quinolone,quinolin-2 1h-one,quinolinol,quinolinone,alpha-quinolone
6038
CHEBI:18289
1H-quinolin-2-one
C1=CC=C2C(=C1)C=CC(=O)N2
Color Brown-Purple or Purple to White
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Melting Point 197.0°C to 200.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 1826
Formula Weight 145.16
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Hydroxyquinoline, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-420-6

RTECSNumber : FG7175000

TSCA : TSCA