Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Imidazolidone hemihydrate, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
10kg
121325-67-5,120-93-4
C6H14N4O3
190.203
MFCD02662355
KPRJGGOOWATRNT-UHFFFAOYSA-N
imidazolidin-2-one hydrate,2-imidazolidinone hemihydrate,2-imidazolidone hemihydrate,bis imidazolidin-2-one hydrate,bis imidazolidinone hydrate,2-imidazolidinone, hydrate 2:1,imidazolidin-2-one, imidazolidin-2-one, hydrate
2723651
imidazolidin-2-one;hydrate
C1CNC(=O)N1.C1CNC(=O)N1.O
121325-67-5,120-93-4
C6H14N4O3
190.203
MFCD02662355
KPRJGGOOWATRNT-UHFFFAOYSA-N
imidazolidin-2-one hydrate,2-imidazolidinone hemihydrate,2-imidazolidone hemihydrate,bis imidazolidin-2-one hydrate,bis imidazolidinone hydrate,2-imidazolidinone, hydrate 2:1,imidazolidin-2-one, imidazolidin-2-one, hydrate
2723651
imidazolidin-2-one;hydrate
C1CNC(=O)N1.C1CNC(=O)N1.O
Boiling Point >100.0°C
Melting Point 58.0°C
Infrared Spectrum Authentic
Quantity 10kg
Color White to Yellow
Density 265g/mL
Flash Point 265°C
Water 13% max.
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Merck Index 14, 4914
Formula Weight 95.1
Physical Form Adhering Crystals or Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material 2-Imidazolidone hemihydrate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Suspected of damaging fertility.

GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning