Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Imino-α-phenyl-3-thiazolidineethanol Hydrochloride 98%, Acros Organics

Click to view available options
Quantity:
25g
15591-41-0
C11H15ClN2OS
258.764
MFCD00044726
HXCSOQNQZVYAJC-UHFFFAOYSA-N
2-2-iminothiazolidin-3-yl-1-phenylethanol hydrochloride,2-2-imino-1,3-thiazolidin-3-yl-1-phenylethan-1-ol hydrochloride,2-imino-alpha-phenyl-3-thiazolidineethanol hydrochloride,2-2-imino-1,3-thiazolidin-3-yl-1-phenylethanol hydrochloride,2-imino-alpha-phenylthiazolidin-3-ethanol monohydrochloride,3-beta-hydroxyphenethyl-2-iminothiazolidine hydrochloride,2-2-imino-1,3-thiazolidin-3-yl-1-phenylethanol,hydrochloride
2723670
2-(2-imino-1,3-thiazolidin-3-yl)-1-phenylethanol;hydrochloride
C1CSC(=N)N1CC(C2=CC=CC=C2)O.Cl
15591-41-0
C11H15ClN2OS
258.764
MFCD00044726
HXCSOQNQZVYAJC-UHFFFAOYSA-N
2-2-iminothiazolidin-3-yl-1-phenylethanol hydrochloride,2-2-imino-1,3-thiazolidin-3-yl-1-phenylethan-1-ol hydrochloride,2-imino-alpha-phenyl-3-thiazolidineethanol hydrochloride,2-2-imino-1,3-thiazolidin-3-yl-1-phenylethanol hydrochloride,2-imino-alpha-phenylthiazolidin-3-ethanol monohydrochloride,3-beta-hydroxyphenethyl-2-iminothiazolidine hydrochloride,2-2-imino-1,3-thiazolidin-3-yl-1-phenylethanol,hydrochloride
2723670
2-(2-imino-1,3-thiazolidin-3-yl)-1-phenylethanol;hydrochloride
C1CSC(=N)N1CC(C2=CC=CC=C2)O.Cl
Quantity 25g
Infrared Spectrum Authentic
Color Beige to White
Melting Point 195°C to 203°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 258.77
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Imino-alpha-phenyl-3-thiazolidineethanol hydrochloride, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.