Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iminopiperidine hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

16011-96-4
C5H11ClN2
134.607
ZHDTXTDHBRADLM-UHFFFAOYSA-N
85236
2,3,4,5-tetrahydropyridin-6-amine;hydrochloride
C1CCN=C(C1)N.Cl

Selective iNOS inhibitor

An inhibitor of NO synthases, more potent than NG-methyl-L-arginine and selective for iNOS.
Formula Weight 134.61
Quantity 50mg
Chemical Name or Material 2-Iminopiperidine hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature