Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Iodo-4-nitrotoluene, 97%

Click to view available options
Quantity:
5g
25g
100g
7745-92-8
C7H6INO2
263.034
MFCD00024329
BUQSRXQJUZTIEW-UHFFFAOYSA-N
2-iodo-4-nitrotoluene,benzene, 2-iodo-1-methyl-4-nitro,2-iodo-1-methyl-4-nitro-benzene,pubchem3957,acmc-209pa2,toluene, 2-iodo-4-nitro,timtec-bb sbb005821
82188
2-iodo-1-methyl-4-nitrobenzene
CC1=C(C=C(C=C1)[N+](=O)[O-])I
7745-92-8
C7H6INO2
263.034
MFCD00024329
BUQSRXQJUZTIEW-UHFFFAOYSA-N
2-iodo-4-nitrotoluene,benzene, 2-iodo-1-methyl-4-nitro,2-iodo-1-methyl-4-nitro-benzene,pubchem3957,acmc-209pa2,toluene, 2-iodo-4-nitro,timtec-bb sbb005821
82188
2-iodo-1-methyl-4-nitrobenzene
CC1=C(C=C(C=C1)[N+](=O)[O-])I
Applications
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.
Quantity 100g
Formula Weight 263.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-4-nitrotoluene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning