Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Iodo-4-(trifluoromethyl)aniline, 99%

Click to view available options
Quantity:
10g
50g
250g
163444-17-5
C7H5F3IN
287.024
MFCD02683097
UKKWTZPXYIYONW-UHFFFAOYSA-N
4-amino-3-iodobenzotrifluoride,2-iodo-4-trifluoromethyl aniline,4-trifluoromethyl-2-iodobenzenamine,benzenamine, 2-iodo-4-trifluoromethyl,2-iodo-4-trifluoromethyl benzenamine,pubchem1632,acmc-209dpp,intermediates-zcf02228,ksc910s3b,3-iodo-4-aminobenzotrifluoride
2783322
2-iodo-4-(trifluoromethyl)aniline
C1=CC(=C(C=C1C(F)(F)F)I)N
163444-17-5
C7H5F3IN
287.024
MFCD02683097
UKKWTZPXYIYONW-UHFFFAOYSA-N
4-amino-3-iodobenzotrifluoride,2-iodo-4-trifluoromethyl aniline,4-trifluoromethyl-2-iodobenzenamine,benzenamine, 2-iodo-4-trifluoromethyl,2-iodo-4-trifluoromethyl benzenamine,pubchem1632,acmc-209dpp,intermediates-zcf02228,ksc910s3b,3-iodo-4-aminobenzotrifluoride
2783322
2-iodo-4-(trifluoromethyl)aniline
C1=CC(=C(C=C1C(F)(F)F)I)N
Melting Point 49°C to 52°C
Quantity 10g
Formula Weight 287.02
Percent Purity 99%
Chemical Name or Material 2-Iodo-4-(trifluoromethyl)aniline

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger