Alfa Aesar™ 2-Iodo-4-(trifluoromethyl)phenol, 97%

Manufacturer: Alfa Aesar™ H33928MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33928MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 463976-21-8
    Molecular Formula C7H4F3IO
    Formula Weight 288.01
    MDL Number MFCD12922960
    UN Number UN3264
    Chemical Name or Material 2-Iodo-4-(trifluoromethyl)phenol

    Specifications

    Formula Weight 288.01
    Molecular Formula C7H4F3IO
    UN Number UN3264
    Quantity 250mg
    MDL Number MFCD12922960
    CAS 463976-21-8
    Boiling Point 105°C (44mmHg)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material 2-Iodo-4-(trifluoromethyl)phenol
    Assay 97%

    Description

    Applications
    2-Iodo-4-(trifluoromethyl)phenol is an pharmaceutical intermediate and it is an intermediate in organic synthesis.

    Notes
    Light sensitive. Store in the dark. Store away from oxidizing agent, light.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit