Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
88-67-5
C7H5IO2
248.01
CJNZAXGUTKBIHP-UHFFFAOYSA-N
o-iodobenzoic acid,benzoic acid, 2-iodo,iodobenzoic acid,benzoic acid, o-iodo,usaf ek-572,2-iodo benzoic acid,2-iodo-benzoic acid,benzoic acid, iodo,kyselina o-jodbenzoova,o-jod-benzoesaeure
6941
CHEBI:287979
2-iodobenzoic acid
C1=CC=C(C(=C1)C(=O)O)I
88-67-5
C7H5IO2
248.01
CJNZAXGUTKBIHP-UHFFFAOYSA-N
o-iodobenzoic acid,benzoic acid, 2-iodo,iodobenzoic acid,benzoic acid, o-iodo,usaf ek-572,2-iodo benzoic acid,2-iodo-benzoic acid,benzoic acid, iodo,kyselina o-jodbenzoova,o-jod-benzoesaeure
6941
CHEBI:287979
2-iodobenzoic acid
C1=CC=C(C(=C1)C(=O)O)I
Color Brown-Orange to Yellow
Packaging Plastic bottle
Melting Point 161.0°C to 163.0°C
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula IC6H4CO2H
Beilstein 09,363
Fieser 01,506
Merck Index 15,5074
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 248.01
Physical Form Powder, Crystals or Flakes
Percent Purity 98%
Chemical Name or Material 2-Iodobenzoic acid
Show More Show Less