Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
5159-41-1
C7H7IO
234.04
MFCD00004610
WZCXOBMFBKSSFA-UHFFFAOYSA-N
2-iodobenzyl alcohol,2-iodophenyl methanol,o-iodobenzyl alcohol,2-iodobenzylalcohol,benzenemethanol, 2-iodo,2-iodobenzylic alcohol,2-jod-benzylalkohol,o-iodo benzylalcohol,2-iodo-benzyl alcohol
107629
(2-iodophenyl)methanol
C1=CC=C(C(=C1)CO)I
5159-41-1
C7H7IO
234.04
MFCD00004610
WZCXOBMFBKSSFA-UHFFFAOYSA-N
2-iodobenzyl alcohol,2-iodophenyl methanol,o-iodobenzyl alcohol,2-iodobenzylalcohol,benzenemethanol, 2-iodo,2-iodobenzylic alcohol,2-jod-benzylalkohol,o-iodo benzylalcohol,2-iodo-benzyl alcohol
107629
(2-iodophenyl)methanol
C1=CC=C(C(=C1)CO)I
Color Pink or White to Yellow
Melting Point 88.0°C to 93.0°C
Infrared Spectrum Authentic
Quantity 10g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula IC6H4CH2OH
Beilstein 06, II, 424
Formula Weight 234.04
Physical Form Needle-Like Powder
Percent Purity 99%
Chemical Name or Material 2-Iodobenzyl alcohol
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-933-2