Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Iodoethanol, 99%, stab. with copper

Click to view available options
Quantity:
5g
25g
100g
624-76-0
C2H5IO
171.965
MFCD00002830
QSECPQCFCWVBKM-UHFFFAOYSA-N
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
12225
2-iodoethanol
C(CI)O
624-76-0
C2H5IO
171.965
MFCD00002830
QSECPQCFCWVBKM-UHFFFAOYSA-N
ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
12225
2-iodoethanol
C(CI)O
Applications
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
Refractive Index 1.572
Sensitivity Light sensitive
Density 2.202
Boiling Point 85°C (25mmHg)
Flash Point 65°C (149°F)
Quantity 5g
Odor Characteristic
Linear Formula ICH2CH2OH
UN Number UN2810
Beilstein 1098258
Solubility Information Soluble in water.
Formula Weight 171.96
Percent Purity 99%
Chemical Name or Material 2-Iodoethanol, stabilized with copper
Show More Show Less

GHS H Statement
H300-H310-H341-H227
Fatal if swallowed.
Fatal in contact with skin.
Suspected of causing genetic defects.
Combustible liquid.

P210-P264b-P270-P280-P301+P310-P330-P370+P378q-P501c

H227-H300

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 210-861-6

RTECSNumber : KL3820000

TSCA : Yes

Recommended Storage : Keep cold