Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5029-67-4
C5H4IN
204.99
MFCD00464928
CCZWSTFVHJPCEM-UHFFFAOYSA-N
2-iodo-pyridine,pyridine, iodo,iodopyridine,2-iodo pyridine,2-iodopyridine,2-pyridyl iodide,2-pyridyl iodide;,pyridine, 2-iodo,pubchem4074,acmc-1avou
221126
2-iodopyridine
C1=CC=NC(=C1)I
5029-67-4
C5H4IN
204.99
MFCD00464928
CCZWSTFVHJPCEM-UHFFFAOYSA-N
2-iodo-pyridine,pyridine, iodo,iodopyridine,2-iodo pyridine,2-iodopyridine,2-pyridyl iodide,2-pyridyl iodide;,pyridine, 2-iodo,pubchem4074,acmc-1avou
221126
2-iodopyridine
C1=CC=NC(=C1)I
Specific Gravity 1.928
Packaging Glass bottle
Boiling Point 52.0°C (0.9 mmHg)
Quantity 5g
Density 1.9280g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 204.99
Percent Purity 98%
Chemical Name or Material 2-Iodopyridine
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning