Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Iodotoluene, 98%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Sensitivity | Light sensitive |
Refractive Index | 1.609 |
Density | 1.713 |
Quantity | 25g |
Flash Point | 90°C (194°F) |
Boiling Point | 210°C to 212°C |
Beilstein | 1634294 |
Formula Weight | 218.04 |
Percent Purity | 98% |
Chemical Name or Material | 2-Iodotoluene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 210-422-9
TSCA : Yes
Recommended Storage : Ambient temperatures