Alfa Aesar™ 2-Isobutyl-4,5-dimethyl-3-thiazoline, 97%

Manufacturer: Alfa Aesar™ H2637903

Quantity 1g

 View more versions of this product

Catalog No. AAH2637903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 65894-83-9
    Molecular Formula C9H17NS
    Formula Weight 171.3
    MDL Number MFCD01680822
    Synonym 2-(2-Methylpropyl)-4,5-dimethyl-3-thiazoline; 4,5-Dimethyl-2-isobutyl-3-thiazoline
    Chemical Name or Material 2-Isobutyl-4,5-dimethyl-3-thiazoline

    Specifications

    Formula Weight 171.3
    TSCA TSCA
    Molecular Formula C9H17NS
    CAS 65894-83-9
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Boiling Point 208°C
    Flash Point 87°C (189°F)
    Quantity 1g
    MDL Number MFCD01680822
    Synonym 2-(2-Methylpropyl)-4,5-dimethyl-3-thiazoline; 4,5-Dimethyl-2-isobutyl-3-thiazoline
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number XJ6642800
    Chemical Name or Material 2-Isobutyl-4,5-dimethyl-3-thiazoline
    Assay 97%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit