Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Isobutyl-4,5-dimethylthiazole, 99%

Click to view available options
Quantity:
2.5g
10g
53498-32-1
C9H15NS
169.286
MFCD00053136
NSVPHVLZAKJSGV-UHFFFAOYSA-N
2-isobutyl-4,5-dimethylthiazole,geranium thiazole,4,5-dimethyl-2-isobutylthiazole,thiazole, 4,5-dimethyl-2-2-methylpropyl,4,5-dimethyl-2-isobutyl thiazole,unii-5p82vd25zi,4,5-dimethyl-2-2-methylpropyl-1,3-thiazole,acmc-1ak7t,2-iso-butyl-4,5-dimethylthiazole,2-isobutyl-4,5-dimethyl-thiazole
104513
4,5-dimethyl-2-(2-methylpropyl)-1,3-thiazole
CC1=C(SC(=N1)CC(C)C)C
53498-32-1
C9H15NS
169.286
MFCD00053136
NSVPHVLZAKJSGV-UHFFFAOYSA-N
2-isobutyl-4,5-dimethylthiazole,geranium thiazole,4,5-dimethyl-2-isobutylthiazole,thiazole, 4,5-dimethyl-2-2-methylpropyl,4,5-dimethyl-2-isobutyl thiazole,unii-5p82vd25zi,4,5-dimethyl-2-2-methylpropyl-1,3-thiazole,acmc-1ak7t,2-iso-butyl-4,5-dimethylthiazole,2-isobutyl-4,5-dimethyl-thiazole
104513
4,5-dimethyl-2-(2-methylpropyl)-1,3-thiazole
CC1=C(SC(=N1)CC(C)C)C
Quantity 10g
Formula Weight 169.28
Percent Purity 99%
Chemical Name or Material 2-Isobutyl-4,5-dimethylthiazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA