Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Isobutylpyridine, 99%
Click to view available options
Quantity:
5g
25g
Specifications
Quantity | 25g |
Density | 0.899 |
Formula Weight | 135.21 |
Percent Purity | 99% |
Chemical Name or Material | 2-Isobutylpyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
EINECSNumber : 228-606-2
TSCA : No
Recommended Storage : Ambient temperatures