Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Colorless |
Density | 0.9120g/mL |
Boiling Point | 73°C (15.0 mmHg) |
Flash Point | 42°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4080 to 1.41 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and k |
Recommended Storage | Refrigerator +4°C |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction