Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropoxyphenol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4812-20-8
C9H12O2
152.19
MFCD00002184
ZNCUUYCDKVNVJH-UHFFFAOYSA-N
2-isopropoxyphenol,o-isopropoxyphenol,phenol, 2-1-methylethoxy,2-1-methylethoxy phenol,2-propan-2-yloxy phenol,unii-x51p4ue6x7,phenol, 2-isopropoxy,2-propan-2-yl-oxy phenol,isopropyl catechol,caswell no. 508a
20949
CHEBI:38547
2-propan-2-yloxyphenol
CC(C)OC1=CC=CC=C1O
4812-20-8
C9H12O2
152.19
MFCD00002184
ZNCUUYCDKVNVJH-UHFFFAOYSA-N
2-isopropoxyphenol,o-isopropoxyphenol,phenol, 2-1-methylethoxy,2-1-methylethoxy phenol,2-propan-2-yloxy phenol,unii-x51p4ue6x7,phenol, 2-isopropoxy,2-propan-2-yl-oxy phenol,isopropyl catechol,caswell no. 508a
20949
CHEBI:38547
2-propan-2-yloxyphenol
CC(C)OC1=CC=CC=C1O
Specific Gravity 1.03
Density 1.0300g/mL
Color Brown-Red to Yellow
Flash Point 87°C
Packaging Glass bottle
Quantity 25g
Boiling Point 100.0°C to 102.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Refractive Index 1.5140 to 1.5160
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2CHOC6H4OH
Formula Weight 152.19
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Isopropoxyphenol, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-379-1

TSCA : TSCA