Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropoxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
138008-97-6
C9H13BO3
180.01
XDMKBIIRBDPSOE-UHFFFAOYSA-N
2-isopropoxyphenylboronic acid,2-isopropoxyphenyl boronic acid,2-isopoxybenzeneboronic acid,2-isopropoxybenzeneboronic acid,boronic acid, 2-1-methylethoxy phenyl,2-propan-2-yloxy phenylboronic acid,2-isopropoxyphenylboronicacid,2-propan-2-yloxyphenyl boronic acid,akos brn-0366,pubchem13297
5084099
(2-propan-2-yloxyphenyl)boronic acid
B(C1=CC=CC=C1OC(C)C)(O)O
138008-97-6
C9H13BO3
180.01
XDMKBIIRBDPSOE-UHFFFAOYSA-N
2-isopropoxyphenylboronic acid,2-isopropoxyphenyl boronic acid,2-isopoxybenzeneboronic acid,2-isopropoxybenzeneboronic acid,boronic acid, 2-1-methylethoxy phenyl,2-propan-2-yloxy phenylboronic acid,2-isopropoxyphenylboronicacid,2-propan-2-yloxyphenyl boronic acid,akos brn-0366,pubchem13297
5084099
(2-propan-2-yloxyphenyl)boronic acid
B(C1=CC=CC=C1OC(C)C)(O)O
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Boiling Point 199.0°C
Melting Point 39.0°C to 43.0°C
Flash Point 110°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 180.01
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Isopropoxyphenylboronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.

GHS Signal Word: Warning