Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Isopropoxypyridine-5-boronic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
870521-30-5
C8H12BNO3
180.998
MFCD07368870
SGEOBUJTURUUJE-UHFFFAOYSA-N
6-isopropoxypyridin-3-yl boronic acid,2-isoproxypyridine-5-boronic acid,6-isopropoxypyridine-3-boronic acid,6-isopropoxypyridin-3-ylboronic acid,6-isopropoxy-3-pyridyl boronic acid,2-isopropoxypyridine-5-boronic acid,2-isopropoxy-5-pyridineboronic acid,6-propan-2-yloxy pyridin-3-yl boronic acid,6-isoproxypyridine-3-boronic acid,boronic acid, 6-1-methylethoxy-3-pyridinyl
44754864
(6-propan-2-yloxypyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)OC(C)C)(O)O
870521-30-5
C8H12BNO3
180.998
MFCD07368870
SGEOBUJTURUUJE-UHFFFAOYSA-N
6-isopropoxypyridin-3-yl boronic acid,2-isoproxypyridine-5-boronic acid,6-isopropoxypyridine-3-boronic acid,6-isopropoxypyridin-3-ylboronic acid,6-isopropoxy-3-pyridyl boronic acid,2-isopropoxypyridine-5-boronic acid,2-isopropoxy-5-pyridineboronic acid,6-propan-2-yloxy pyridin-3-yl boronic acid,6-isoproxypyridine-3-boronic acid,boronic acid, 6-1-methylethoxy-3-pyridinyl
44754864
(6-propan-2-yloxypyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)OC(C)C)(O)O
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Formula Weight 181
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Isopropoxypyridine-5-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning