Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropyl-4-methylthiazole, 99%, Acros Organics

Click to view available options
Quantity:
5g
15679-13-7
C7H11NS
141.232
MFCD00036561
OFLXNHNYPQPQKW-UHFFFAOYSA-N
2-isopropyl-4-methylthiazole,4-methyl-2-1-methylethyl thiazole,2-isopropyl-4-methyl thiazole,thiazole, 2-isopropyl-4-methyl,thiazole, 4-methyl-2-1-methylethyl,unii-nqf7jo3mup,tropical thiazole,2-isopropyl-4-methyl-1,3-thiazole,fema no. 3555,nqf7jo3mup
61808
4-methyl-2-propan-2-yl-1,3-thiazole
CC1=CSC(=N1)C(C)C
15679-13-7
C7H11NS
141.232
MFCD00036561
OFLXNHNYPQPQKW-UHFFFAOYSA-N
2-isopropyl-4-methylthiazole,4-methyl-2-1-methylethyl thiazole,2-isopropyl-4-methyl thiazole,thiazole, 2-isopropyl-4-methyl,thiazole, 4-methyl-2-1-methylethyl,unii-nqf7jo3mup,tropical thiazole,2-isopropyl-4-methyl-1,3-thiazole,fema no. 3555,nqf7jo3mup
61808
4-methyl-2-propan-2-yl-1,3-thiazole
CC1=CSC(=N1)C(C)C
Color Colorless to Yellow
Quantity 5g
Refractive Index 1.4960 to 1.5010 (20°C, 589nm)
Infrared Spectrum Authentic
Density 0.997g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 141.23
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Isopropyl-4-methylthiazole, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.