Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropyl-5-methylhexyl Acetate 98.0+%, TCI America™

Click to view available options
25ML
40853-55-2
C12H24O2
200.322
MFCD00129077
QLFSGYMLVZGFJT-UHFFFAOYSA-N
Acetic Acid 2-Isopropyl-5-methylhexyl Ester, Tetrahydrolavandulyl Acetate
524263
(5-methyl-2-propan-2-ylhexyl) acetate
CC(C)CCC(COC(=O)C)C(C)C
40853-55-2
C12H24O2
200.322
MFCD00129077
QLFSGYMLVZGFJT-UHFFFAOYSA-N
Acetic Acid 2-Isopropyl-5-methylhexyl Ester, Tetrahydrolavandulyl Acetate
524263
(5-methyl-2-propan-2-ylhexyl) acetate
CC(C)CCC(COC(=O)C)C(C)C
Boiling Point 220°C
Color Colorless
Quantity 25mL
Formula Weight 200.32
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Isopropyl-5-methylhexyl Acetate