Learn More
2-Isopropylaniline, 97%, Thermo Scientific Chemicals
Description
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1?-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1′-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides. Light Sensitive
Specifications
Density | 0.966 |
Boiling Point | 214°C to 216°C |
Flash Point | 95°C (203°F) |
Refractive Index | 1.5485 |
Quantity | 25 g |
Beilstein | 636283 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
IUPAC Name | 2-(propan-2-yl)aniline |
Formula Weight | 135.21 |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H336-H373 |
Hazard Statement | GHS H Statement H311-H373-H302-H332-H315-H319 Toxic in contact with skin. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P363-P501c |
EINECSNumber | 211-397-7 |
RTECSNumber | BY4200000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.