Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Isopropylaniline, 97%

Click to view available options
Quantity:
25g
100g
500g
643-28-7
C9H13N
135.21
MFCD00007720
YKOLZVXSPGIIBJ-UHFFFAOYSA-N
2-isopropylaniline,o-isopropylaniline,o-cumidine,2-propan-2-yl aniline,benzenamine, 2-1-methylethyl,o-aminoisopropylbenzene,2-isopropyl aniline,2-aminoisopropylbenzene,aniline, o-isopropyl,o-isopropyl aniline
12561
2-propan-2-ylaniline
CC(C)C1=CC=CC=C1N
643-28-7
C9H13N
135.21
MFCD00007720
YKOLZVXSPGIIBJ-UHFFFAOYSA-N
2-isopropylaniline,o-isopropylaniline,o-cumidine,2-propan-2-yl aniline,benzenamine, 2-1-methylethyl,o-aminoisopropylbenzene,2-isopropyl aniline,2-aminoisopropylbenzene,aniline, o-isopropyl,o-isopropyl aniline
12561
2-propan-2-ylaniline
CC(C)C1=CC=CC=C1N
Applications
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1′-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides. Light Sensitive
Quantity 25g
Flash Point 95°C (203°F)
Density 0.966
Refractive Index 1.5485
Sensitivity Light sensitive
Boiling Point 214°C to 216°C
Beilstein 636283
Solubility Information Insoluble in water.
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 2-Isopropylaniline
Show More Show Less

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P363-P501c

H302+H312+H332-H336-H373

EINECSNumber : 211-397-7

RTECSNumber : BY4200000

TSCA : Yes

Recommended Storage : Ambient temperatures