Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Isopropylanisole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Density | 0.936 |
Boiling Point | 198°C to 200°C |
Flash Point | 86°C (187°F) |
Refractive Index | 1.508 |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 150.22 |
Percent Purity | 98% |
Chemical Name or Material | 2-Isopropylanisole |
Safety and Handling
Hazard Category | H227 |
Hazard Statement | GHS H Statement H227 Combustible liquid. |
Precautionary Statement | P210-P280-P370+P378q-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction