Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylimidazole, 98%, Thermo Scientific Chemicals

Catalog Number ac442181000
Click to view available options
Quantity:
100 g
36947-68-9
C6H10N2
110.16
MFCD00014486
FUOZJYASZOSONT-UHFFFAOYSA-N
2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole
123457
2-propan-2-yl-1H-imidazole
CC(C)C1=NC=CN1
36947-68-9
C6H10N2
110.16
MFCD00014486
FUOZJYASZOSONT-UHFFFAOYSA-N
2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole
123457
2-propan-2-yl-1H-imidazole
CC(C)C1=NC=CN1

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 130°C to 132°C
Color Brown to White
Boiling Point 163°C to 166°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 g
Formula Weight 110.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Isopropylimidazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-286-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit