2-Isopropylphenol, 98%

Manufacturer: Alfa Aesar A1202730

Quantity 250g

 View more versions of this product

Catalog No. AAA1202730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88-69-7
    Molecular Formula C9H12O
    Formula Weight 136.2
    MDL Number MFCD00002224
    UN Number UN2922
    Beilstein 1363322
    Chemical Name or Material 2-Isopropylphenol

    Specifications

    Formula Weight 136.2
    Quantity 250g
    TSCA TSCA
    MDL Number MFCD00002224
    UN Number UN2922
    Beilstein 1363322
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON S
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    RTECS Number SL5900000
    Density 1.010
    Molecular Formula C9H12O
    CAS 88-69-7
    Assay Percent Range 98%
    Chemical Name or Material 2-Isopropylphenol
    Assay 98%

    GHS H Statement
    H301-H311-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON S

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit