Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylphenyl isocyanate, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
56309-56-9
C10H11NO
161.21
MWBSQVPLYJZPTG-UHFFFAOYSA-N
2-isopropylphenyl isocyanate,1-isocyanato-2-isopropylbenzene,o-isopropylphenyl isocyanate,2-isopropylphenylisocyanate,1-isocyanato-2-propan-2-yl benzene,1-isocyanato-2-isopropyl-benzene,2-methylethyl benzenisocyanate,acmc-1avxp,isopropylphenyl isocyanate
91797
1-isocyanato-2-propan-2-ylbenzene
CC(C)C1=CC=CC=C1N=C=O
56309-56-9
C10H11NO
161.21
MWBSQVPLYJZPTG-UHFFFAOYSA-N
2-isopropylphenyl isocyanate,1-isocyanato-2-isopropylbenzene,o-isopropylphenyl isocyanate,2-isopropylphenylisocyanate,1-isocyanato-2-propan-2-yl benzene,1-isocyanato-2-isopropyl-benzene,2-methylethyl benzenisocyanate,acmc-1avxp,isopropylphenyl isocyanate
91797
1-isocyanato-2-propan-2-ylbenzene
CC(C)C1=CC=CC=C1N=C=O
Quantity 5g
Boiling Point 66.0°C to 67.0°C (5.0mmHg)
Specific Gravity 1.019
Color Colorless
Density 1.0190g/mL
Flash Point 88°C
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 161.21
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-Isopropylphenyl isocyanate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 260-105-4