Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Mercapto-4,5-dimethylthiazole, 95%

Click to view available options
Quantity:
250mg
1g
5g
5351-51-9
C5H7NS2
145.238
MFCD00037895
KKHBRTFQIYIHEI-UHFFFAOYSA-N
4,5-dimethylthiazole-2-thiol,4,5-dimethyl-thiazole-2-thiol,4,5-dimethylthiazole-2 3h-thione,dimethyl-1,3-thiazole-2-thiol,4,5-dimethyl-1,3-thiazole-2-thiol,2 3h-thiazolethione, 4,5-dimethyl,2-thiazolethiol, 4,5-dimethyl,4,5-dimethyl-2-mercaptothiazole,2-mercapto-4,5-dimethylthiazole,4,5-dimethyl-2-thiazolethiol
817026
4,5-dimethyl-3H-1,3-thiazole-2-thione
CC1=C(SC(=S)N1)C
5351-51-9
C5H7NS2
145.238
MFCD00037895
KKHBRTFQIYIHEI-UHFFFAOYSA-N
4,5-dimethylthiazole-2-thiol,4,5-dimethyl-thiazole-2-thiol,4,5-dimethylthiazole-2 3h-thione,dimethyl-1,3-thiazole-2-thiol,4,5-dimethyl-1,3-thiazole-2-thiol,2 3h-thiazolethione, 4,5-dimethyl,2-thiazolethiol, 4,5-dimethyl,4,5-dimethyl-2-mercaptothiazole,2-mercapto-4,5-dimethylthiazole,4,5-dimethyl-2-thiazolethiol
817026
4,5-dimethyl-3H-1,3-thiazole-2-thione
CC1=C(SC(=S)N1)C
Quantity 250mg
Beilstein 107610
Formula Weight 145.24
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Mercapto-4,5-dimethylthiazole