Alfa Aesar™ 2-Mercapto-4,6-dimethylpyrimidine hydrate, 98%

Manufacturer: Alfa Aesar™ A1223109

Quantity 10g

 View more versions of this product

Catalog No. AAA1223109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22325-27-5
    Molecular Formula C6H8N2S·xH2O
    Formula Weight 140.21 (Anhydrous)
    MDL Number MFCD00006079
    Beilstein 81330
    Synonym 4,6-Dimethylpyrimidine-2-thiol
    Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine hydrate

    Specifications

    Formula Weight 140.21 (Anhydrous)
    Molecular Formula C6H8N2S·xH2O
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 10g
    MDL Number MFCD00006079
    CAS 22325-27-5
    Synonym 4,6-Dimethylpyrimidine-2-thiol
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Melting Point 213° to 216°C
    Beilstein 81330
    RTECS Number UW5650000
    Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine hydrate
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit