Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-4-methylpyrimidine hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
6959-66-6
C5H6N2S·HCl
162.64
MFCD00012783
UQJLPBLXSJWAKG-UHFFFAOYSA-N
2-mercapto-4-methylpyrimidine hydrochloride,4-methylpyrimidine-2-thiol hydrochloride,4-methylpyrimidine-2 1h-thione hydrochloride,6-methyl-1,2-dihydropyrimidine-2-thione hydrochloride,4-methylpyrimidine-2-thiol hcl,2 1h-pyrimidinethione, 4-methyl-, monohydrochloride,c5h6n2s.clh,2-mercapto-4-methyl pyrimidine hydrochloride,pubchem21990,acmc-1b3ky
2723697
6-methyl-1H-pyrimidine-2-thione;hydrochloride
CC1=CC=NC(=S)N1.Cl
6959-66-6
C5H6N2S·HCl
162.64
MFCD00012783
UQJLPBLXSJWAKG-UHFFFAOYSA-N
2-mercapto-4-methylpyrimidine hydrochloride,4-methylpyrimidine-2-thiol hydrochloride,4-methylpyrimidine-2 1h-thione hydrochloride,6-methyl-1,2-dihydropyrimidine-2-thione hydrochloride,4-methylpyrimidine-2-thiol hcl,2 1h-pyrimidinethione, 4-methyl-, monohydrochloride,c5h6n2s.clh,2-mercapto-4-methyl pyrimidine hydrochloride,pubchem21990,acmc-1b3ky
2723697
6-methyl-1H-pyrimidine-2-thione;hydrochloride
CC1=CC=NC(=S)N1.Cl
Color Yellow
Quantity 500g
Infrared Spectrum Authentic
Melting Point 265.0°C
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 162.64
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2-Mercapto-4-methylpyrimidine hydrochloride
Show More Show Less